Learn More
Eppendorf™ Heat Sealer Film and Foils
GREENER_CHOICE
Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination
Brand:  Eppendorf™ 0030127838
 View more versions of this product
Product Code. 11833432
Quantity Price
1 £183.00 / Pack of 100
5 £160.89 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
  • Transparent heat sealing film allows sample monitoring in the plates
  • Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
  • Foil stripper tool removes heat-sealed film and foils completely
  • Certified free of DNA, DNase, RNase, and PCR inhibitors
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Autoclavable
Hermetic sealing of PCR plates, especially recommended for low reaction volumes, best protection against evaporation
Yes
Polyester, Polypropylene
100
PCR Film
-80°C to +100°C