Learn More
Eppendorf™ Reference™ 2, Mechanical Single-Channel Pipettes Multi-Packs
GREENER_CHOICE
Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorf™ Reference™ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants. Available in packs of three, four and six.
£810.00 - £1767.04
Specifications
Autoclavable Autoclavable
Number of Channels 1
View More Specs
Products 4
Product Code Brand Includes Volume (Metric) Price Quantity & Availability  
Product Code Brand Includes Volume (Metric) Price Quantity & Availability  
15763499
GREENER_CHOICE
View Documents
Eppendorf™
4924000916
3 Pipettes: Reference™ 2 1-Channel Variable, 3 x epT.I.P.S.™ Box 2.0 with 96 tips, Ballpoint Pen 2 to 20, 20 to 200, 100 to 1000 μL
£813.24
1 set
In Stock
to see stock.
Add to basket
15753499
GREENER_CHOICE
View Documents
Eppendorf™
4924000908
3 Pipettes: Reference™ 2 1-Channel Variable, 3 x epT.I.P.S.™ Box 2.0 with 96 tips, Ballpoint Pen 0.5 to 10, 10 to 100, 100 to 1000 μL
£813.24
1 set
In Stock
to see stock.
Add to basket
15773499
GREENER_CHOICE
View Documents
Eppendorf™
4924000924
3 Pipettes: Reference™ 2 1-Channel Variable, 1 x epT.I.P.S.™ Box 2.0, 2 x epT.I.P.S. sample bags, Ballpoint Pen 100 to 1000 μL, 0.5 to 5 mL, 1 to 10 mL
£810.00
1 set
In Stock
to see stock.
Add to basket
17894601
View Documents Promotion Details
Eppendorf™
4924000940
6 Pipettes: Reference™ 2 1-Channel Variable, 1 x Pipette Carousel 2, 6 x epT.I.P.S.™ Box 2.0 with 96 tips, Ballpoint Pen 0.1 to 2.5, 0.5 to 10, 2 to 20, 10 to 100, 20 to 200, 100 to 1000 μL
£1767.04
1 set
In Stock
to see stock.
Add to basket
Description
Description
  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
Specifications
Specifications
Autoclavable
1