Learn More
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume
GREENER_CHOICE
Resistant to chemical, thermal, and mechanical stress
£170.68 - £186.91
Specifications
Autoclavable Autoclavable
Type Fixed Volume Pipette
View More Specs
Products 12
Product Code Brand Volume (Metric) Accuracy Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Accuracy Color Price Quantity & Availability  
15753459
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000016
10 μL ±1.2% Gray
£186.91
Each
In Stock
to see stock.
Add to basket
15763459
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000024
10 μL ±1.2% Yellow
£179.72
Each
In Stock
to see stock.
Add to basket
15773459
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000032
20 μL ±0.8% Gray
£179.72
Each
In Stock
to see stock.
Add to basket
15783459
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000040
20 μL ±1% Yellow
£179.72
Each
In Stock
to see stock.
Add to basket
15793459
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000059
25 μL ±1% Yellow
£186.91
Each
In Stock
to see stock.
Add to basket
15703469
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000067
50 μL ±0.7% Yellow
£186.91
Each
In Stock
to see stock.
Add to basket
15713469
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000075
100 μL ±0.6% Yellow
£186.91
Each
In Stock
to see stock.
Add to basket
15733469
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000091
200 μL ±0.6% Blue
£170.68
Each
In Stock
to see stock.
Add to basket
15723469
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000083
200 μL ±0.6% Yellow
£179.72
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
15743469
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000105
250 μL ±0.6% Blue
£170.68
Each
In Stock
to see stock.
Add to basket
15753469
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000113
500 μL ±0.6% Blue
£186.91
Each
In Stock
to see stock.
Add to basket
15763469
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
3124000121
1000 μL ±0.6% Blue
£186.91
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume
Specifications
Specifications
Autoclavable
Fixed Volume Pipette
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume