missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
Brand: Eppendorf™ 5409714007
538.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.

Specifications
F-45-48-5-PCR | |
48 x 0.2 mL PCR Tubes | |
10,500 rpm | |
Plastic Rotor Lid | |
F-45-48-5-PCR |
Rotor | |
11,710 x g | |
Centrifuge 5427 R | |
48 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ Rotor for Centrifuge 5427 R > Description: F-45-48-5-PCR, Capacity: 48 x 0.2 mL PCR Tubes, Max. RCF: 11,710 x g, Max. Speed: 10,500 rpm
Spot an opportunity for improvement?Share a Content Correction