Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
Brand:  Eppendorf™ 5409702009
666.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 13443396
  • £702.80 / Each
Explore more special offers
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FA-45-24-11, Aerosol-tight - includes rotor lid
24 x 1.5 to 2.0 mL Tubes
16,220 rpm
Rotor Lid
Aerosol-tight Rotor
25,001 x g
Centrifuge 5427 R
FA-45-24-11
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Rotor for Centrifuge 5427 R > Description: FA-45-24-11, Aerosol-tight - includes rotor lid, Capacity: 24 x 1.5 to 2.0 mL Tubes, Max. RCF: 25,001 x g, Max. Speed: 16,220 rpm