Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
Brand:  Eppendorf™ 5409708007
427.95 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 13463396
  • £445.78 / Each
Explore more special offers
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
F-45-30-11 - includes rotor lid
30 x 1.5 to 2.0 mL Tubes
14,000 rpm
Rotor Lid
F-45-30-11
Rotor
20,817 x g
Centrifuge 5427 R
30
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Rotor for Centrifuge 5427 R > Description: F-45-30-11 - includes rotor lid, Capacity: 30 x 1.5 to 2.0 mL Tubes, Max. RCF: 20,817 x g, Max. Speed: 14,000 rpm