Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
Brand:  Eppendorf™ 5409704001
735.86 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 13483396
  • £802.00 / Each
Explore more special offers
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FA-45-24-11-Kit, Aerosol-tight - includes rotor lid
24 Spin Columns
13,200 rpm
Rotor Lid
FA-45-24-11-Kit
Aerosol-tight Rotor
19,090 x g
Centrifuge 5427 R
24
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Rotor for Centrifuge 5427 R > Description: FA-45-24-11-Kit, Aerosol-tight - includes rotor lid, Capacity: 24 Spin Columns, Max. RCF: 19,090 x g, Max. Speed: 13,200 rpm