missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube
GREENER_CHOICE

Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube ensures secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1.5 mL capacity.

£30.49

Specifications

Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Material Polypropylene
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
View More Specs

Products 1
Product Code Brand Color Purity or Quality Grade Max. RCF Price Quantity & Availability  
Product Code Brand Color Purity or Quality Grade Max. RCF Price Quantity & Availability  
15625367
GREENER_CHOICE
View Documents
Eppendorf™
0030123611
Clear Forensic DNA Grade 22,000 x g
£30.49
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • The hinged Eppendorf Safe-Lock lid prevents inadvertent lid opening during incubation and storage.
  • g-Safe™ technology, depending on the Safe-Lock Tube variant (refer to the manual), ensures outstanding centrifugation stability up to 30,000 x g, preventing sample loss due to tube breakage and providing additional safety when handling hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances demonstrated to leach from plastic consumables into the sample and adversely impact bio-assay results.
  • Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures.
  • Precision lid sealing minimizes evaporation rates during extended storage periods.
  • The generously sized frosted lid and side surface facilitate easy labeling.
  • Assured functionality in a temperature range from -86 °C to 100 °C.
  • Autoclavable when open (121 °C, 20 min) for reliable sterilization.
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur™ quality for tested purity.

Order Info

Supplied individually wrapped in your choice of three sizes

Specifications

Specifications

Autoclavable
Polypropylene
-86°C to +100°C
0.05 oz.
Cylindrical
1.5 mL
-122.8°F to +212°F
Yes
Safe-lock Lid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.