missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals EPPS, 99%, for biochemistry

£82.71 - £276.38

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym 3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10104480
View Documents Promotion Details
Thermo Scientific Chemicals
172601000
100 g Glass bottle
£82.71
100g
Please to purchase this item. Need a web account? Register with us today!
10474205
View Documents Promotion Details
Thermo Scientific Chemicals
172605000
500 g Glass bottle
£276.38
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

16052-06-5
252.329
OWXMKDGYPWMGEB-UHFFFAOYSA-N
85255
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C9H20N2O4S
MFCD00006160
3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
CHEBI:42298
C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

(0.1 M aq. soln.) (1 cm cell)
16052-06-5
5 to 6 (10% w/v soln. at 20°C)
99%
C9H20N2O4S
3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
C1CN(CCN1CCCS(=O)(=O)O)CCO
252.329
CHEBI:42298
≥98.5%
EPPS
(0.1M aq. soln.) (1 cm cell)
White
Crystalline Powder
Authentic
MFCD00006160
OWXMKDGYPWMGEB-UHFFFAOYSA-N
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
85255
252.33
Biochemical
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • EPPS

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only