missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPRS Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | EPRS |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803905
|
Proteintech
25307-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16893895
|
Proteintech
25307-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EPRS is a member of aminoacyl-tRNA synthetases, a class of enzymes that charge tRNAs with their cognate amino acids. EPRS is a multifunctional aminoacyl-tRNA synthetase that catalyzes the aminoacylation of glutamic acid and proline tRNA species.Specifications
EPRS | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
P07814 | |
2058 | |
EPRS Fusion Protein Ag18011 | |
Primary | |
-20°C | |
EPRS |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EARS, EPRS, GLNS, GLUPRORS, GluRS Prolyl tRNA synthetase, Glutamate tRNA ligase, PARS, PIG32, Proline tRNA ligase, QARS, QPRS | |
EPRS | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |