missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactam, 99%

Product Code. p-7044536
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 11357086

Brand: Thermo Scientific Alfa Aesar L06999.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-60-2 | C6H11NO | 113.16 g/mol

ε-Caprolactam is used as a monomer in the preparation of synthetic fibers of the polyamide as well as a solvent for high molecular weight polymers. It is also used in coatings and plasticizers. Further, it is used as a cross linking agent for polyurethanes. In addition to this, it is used to prepare nylon-6, which finds application in fibers and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ε-Caprolactam is used as a monomer in the preparation of synthetic fibers of the polyamide as well as a solvent for high molecular weight polymers. It is also used in coatings and plasticizers. Further, it is used as a cross linking agent for polyurethanes. In addition to this, it is used to prepare nylon-6, which finds application in fibers and plastics.

Solubility
Soluble in benzene, ethanol, water, chloroform, methanol, tetrahydrofurfuryl alcohol, ether, dimethylformamide, chlorinated hydrocarbons, cyclohexene and petroleum fractions.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-60-2
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006936
InChI Key JBKVHLHDHHXQEQ-UHFFFAOYSA-N
Synonym epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam
PubChem CID 7768
ChEBI CHEBI:28579
IUPAC Name azepan-2-one
SMILES C1CCC(=O)NCC1

Specifications

Melting Point 68°C to 72°C
Density 1.01
pH 7 to 8.5
Boiling Point 270°C
Flash Point 152°C (305°F)
Quantity 100 g
Beilstein 106934
Merck Index 14,1761
Solubility Information Soluble in benzene,ethanol,water,chloroform,methanol,tetrahydrofurfuryl alcohol,ether,dimethylformamide,chlorinated hydrocarbons,cyclohexene and petroleum fractions.
Formula Weight 113.16
Percent Purity 99%
Chemical Name or Material epsilon-Caprolactam
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.