missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ epTIPS™ Low-retention Pipettor Tip Box
Colour coded box for storing epTIPS pipettor tips
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With (Application) | 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack |
Format | Rack |
Tip Style | Low Retention |
Description
Colour coded box for storing epTIPS pipettor tips
- Contamination free transfer of trays to the refill box (epTIPS box) in plastic covers
- Racks can be autoclaved
- Also available in PCR clean quality, free of human DNA, DNase, RNase and PCR inhibitors
- 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
Autoclavable | |
0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack | |
Rack | |
Low Retention |