Learn More
Eppendorf™ epTIPS™ Low-retention Pipettor Tips
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tip Reloads are ideal for cell culture (media) applications and applications with samples that contain detergents.
Specifications
Sterility Sterile
For Use With (Application) They satisfy the most stringent demands in medicine, the pharmaceutical and food industries, PCR*, molecular biology and cytotechnology
Compatibility Eppendorf™
Format Refill
Tip Style Low Retention
View More Specs
Description
Description
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tip Reloads are ideal for cell culture (media) applications and applications with samples that contain detergents.
  • Ultrahomogenous surface for maximum reproducibility
  • Ultrahydrophobic surface for minimal sample loss
  • Significantly reduced foam formation during pipetting
  • Extreme resistance to chemicals
  • Available in PCR clean and Eppendorf Quality
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free)
  • No coating
Specifications
Specifications
Sterile
Eppendorf™
Low Retention
They satisfy the most stringent demands in medicine, the pharmaceutical and food industries, PCR*, molecular biology and cytotechnology
Refill
Eppendorf™ Pipettor
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Eppendorf™ epTIPS™ Low-retention Pipettor Tips