missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Equine IgG Isotype Control, Agarose, Novus Biologicals™
Horse Antibody
Brand: Bio-Techne NBP1-97139
Additional Details : Weight : 0.00970kg
Description
IgG specifically detects IgG in Equine samples. It is validated for Immunoprecipitation.Specifications
IgG | |
Agarose | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide | |
IGHG1 | |
Multi-step | |
RUO | |
3500 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunoprecipitation | |
Immunoprecipitation 1:10 - 1:500 | |
Immunoglobulin G, ImmunoglobulinG | |
Equine | |
10 mg | |
Isotype Control (Primary) | |
Equine | |
IgG |
For Research Use Only