Learn More
Thermo Scientific™ EquiPhi29™ DNA Amplification Kit
The Thermo Scientific EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification – whole genome amplification (MDA-WGA).
£321.00 - £1338.00
Specifications
Product Type | DNA Amplification Kit |
---|---|
Form | Liquid |
For Use With (Application) | RCA, MDA-WGA |
Polymerase | EquiPhi29 DNA Polymerase |
Research Category | Nucleic Acid Analysis, In Vitro Transcription, Cell-free Protein Expression, In Situ Sequencing, SNP Detection |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18094070
|
Thermo Scientific™
A65393 |
100 Reactions |
£321.00
100 reactions |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18084100
|
Thermo Scientific™
A65394 |
500 Reactions |
£1338.00
500 reactions |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Thermo Scientific EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification – whole genome amplification (MDA-WGA).
Features of the EquiPhi29 DNA Amplification Kit include:
• Fast DNA amplification in less than two hours
• Sensitivity from 1 fg of input DNA
• Maximal yield of 17 µg amplified DNA
• Compatibility with different types of sample input material (e.g., purified DNA, liquid media culture, agar plate colonies)
• Compatibility with various downstream applications such as DNA sequencing (Sanger sequencing, next-generation sequencing), digestion with restriction enzymes, cell-free DNA enrichment, and cell-free protein expression
• All required components for DNA amplification in RCA or MDA-WGA reactions
The kit contains EquPhi29 DNA Polymerase, a proprietary phi29 DNA polymerase mutant that possesses strong strand displacement activity and enables high thermostability, reaction speed, and product yield and low amplification bias. Also included is an optimized reaction buffer, exo-resistant random primer, pyrophosphatase, and dNTPs. The addition of pyrophosphatase to the reaction mixture enhances DNA amplification and facilitates sample handling.
Specifications
DNA Amplification Kit | |
RCA, MDA-WGA | |
Nucleic Acid Analysis, In Vitro Transcription, Cell-free Protein Expression, In Situ Sequencing, SNP Detection | |
42°C | |
Up to 2 hours |
Liquid | |
EquiPhi29 DNA Polymerase | |
Kit | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.