missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2 (HER-2), His Tag ErbB2 (HER2), His Tag Recombinant Protein
ErbB2 (HER-2), His Tag ErbB2 (HER2), His Tag Recombinant Protein
Brand: Invitrogen PV3366
This item is not returnable.
View return policy
Description
ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.Specifications
NP_004439 | |
2064 | |
10μg | |
EGF Receptor (ErbB) Kinase Family | |
Human | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% CHAPS, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™ | |
V-erb-b2 erythroblastic leukemia viral oncogene homolog 2 | |
Kinases & Inhibitors | |
Receptor Tyrosine Kinases |
Kinase Assay | |
68.6 kDa | |
ErbB2 (HER2), NEU | |
aa 676-1255 | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity | |
Dry Ice | |
ERBB2 | |
Baculovirus | |
Recombinant, Cytoplasmic | |
Oncology, Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.