missing translation for 'onlineSavingsMsg'
Learn More
Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11321779

Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11321779
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11321779

Brand: Thermo Scientific Alfa Aesar 012388.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Erbium foil used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films. It is used as an alloying element with titanium and is also used to produce infra-red absorbing glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]

Specifications

UN Number UN3178
Quantity 25 x 25 mm
Solubility Information Insoluble in water.
Percent Purity 99.9%
Weight ≈0.14g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Erbium foil, 0.025mm (0.001 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP29J042 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number012388.FF
Lot NumberB22X025 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012388.FF
Lot NumberA08T019 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012388.FF
Lot NumberY26F030 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012388.FF
Lot NumberA17Q24 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012388.FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.