missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erbium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals
£105.00 - £401.00
Chemical Identifiers
CAS | 7440-52-0 |
---|---|
Molecular Formula | Er |
Molecular Weight (g/mol) | 167.26 |
MDL Number | MFCD00010987 |
InChI Key | UYAHIZSMUZPPFV-UHFFFAOYSA-N |
Synonym | unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic |
PubChem CID | 23980 |
ChEBI | CHEBI:33379 |
IUPAC Name | erbium |
SMILES | [Er] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311779
|
Thermo Scientific Alfa Aesar
012386.FF |
25 x 25 mm |
£105.00
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460270
|
Thermo Scientific Alfa Aesar
012386.FI |
50 x 50 mm |
£401.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-52-0 | |
167.26 | |
UYAHIZSMUZPPFV-UHFFFAOYSA-N | |
23980 | |
erbium |
Er | |
MFCD00010987 | |
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic | |
CHEBI:33379 | |
[Er] |
Specifications
7440-52-0 | |
MFCD00010987 | |
UYAHIZSMUZPPFV-UHFFFAOYSA-N | |
erbium | |
23980 | |
99.9% | |
≈1.4g/25x25mm | |
(REO) |
Er | |
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic | |
[Er] | |
167.26 | |
CHEBI:33379 | |
Odorless | |
Foil | |
Erbium foil, 0.25mm (0.01 in.) thick |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'einecsNumber' : 231-160-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only