missing translation for 'onlineSavingsMsg'
Learn More
Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11366668

Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11366668
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11366668

Brand: Thermo Scientific Alfa Aesar 013214.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Erbium(III) 2,4-pentanedionate is commonly used in various catalysts and catalytic reagents for organic synthesis. It is also used for glass coloring, as an amplifier in fiber optics, and in lasers for medical and dental purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14553-08-3
Molecular Formula C15H21ErO6
Molecular Weight (g/mol) 464.59
MDL Number MFCD00013490
InChI Key FGQSJRDKBCVFHH-UHFFFAOYSA-N
Synonym erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-oneShow MoreShow Less
PubChem CID 102601666
SMILES [Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
IUPAC Name erbium;(Z)-4-hydroxypent-3-en-2-one
Formula Weight 464.59
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Erbium(III) 2, 4-pentanedionate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM11I023 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number013214.04,013214.09
Lot NumberC25T038 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013214.04,013214.09
Lot NumberQ03G004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013214.04,013214.09
Lot NumberF05L44 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013214.04,013214.09
Lot NumberF21X010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013214.04,013214.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.