missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erbium(III) acetate tetrahydrate, 99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Erbium(III) acetate tetrahydrate is used for laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components, photo-optical material and optical fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Pink |
Physical Form | Crystalline Aggregates |
Quantity | 100 g |
Linear Formula | Er(OOCCH3)3·4H2O |
Solubility Information | Soluble in water. |
Formula Weight | 416.45 (344.39 Anhydrous) |
Percent Purity | 99% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 2.114 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'