missing translation for 'onlineSavingsMsg'
Learn More
Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11388458

Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11388458
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11388458

Brand: Thermo Scientific Alfa Aesar 040481.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Erbium(III) acetate tetrahydrate participates in the synthesis and determination of crystal structures and stereochemical properties of lanthanide(III) complexes with ethylenediamine-N,N,N?,N?-tetraacetate. Participates in simple one-pot single-step synthesis of rare earth doped spherical mesoporous metal oxide nanospheres with upconversion emission ability in supercritical methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15280-57-6
MDL Number MFCD00013034

Specifications

Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula Er(OOCCH3)3·4H2O
Solubility Information Soluble in water.
Formula Weight 416.45 (344.39 Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 2.114 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) acetate tetrahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ30K009 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number040481.14,040481.06,040481.22
Lot NumberX27H019 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040481.14,040481.06,040481.22
Lot NumberF21L31 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040481.14,040481.06,040481.22
Lot NumberU16H033 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040481.14,040481.06,040481.22
Lot NumberN07F028 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040481.14,040481.06,040481.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.