missing translation for 'onlineSavingsMsg'
Learn More
Erbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11351049

Erbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11351049
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11351049

Brand: Thermo Scientific Alfa Aesar 011304.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Erbium(III) chloride hydrate is used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18, for which electron and stability studies were performed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-85-9
Molecular Formula Cl3Er
Molecular Weight (g/mol) 273.61
MDL Number MFCD00010989
InChI Key HDGGAKOVUDZYES-UHFFFAOYSA-K
IUPAC Name erbium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Er+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 25 g
Solubility Information Slightly soluble in ethanol
Formula Weight 273.62 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) chloride hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM08H036 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number011304.22,011304.14
Lot NumberB08Q23A Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011304.22,011304.14
Lot NumberI20X015 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011304.22,011304.14
Lot NumberF12L22 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011304.22,011304.14
Lot NumberL21Y070 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011304.22,011304.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.