Learn More
Erbium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
£155.78 - £526.62
Chemical Identifiers
CAS | 19423-85-9 |
---|---|
Molecular Formula | Cl3Er |
Molecular Weight (g/mol) | 273.61 |
MDL Number | MFCD00010989 |
InChI Key | HDGGAKOVUDZYES-UHFFFAOYSA-K |
IUPAC Name | erbium(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Er+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371049
|
Thermo Scientific Alfa Aesar
011305.14 |
25 g |
£155.78
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361049
|
Thermo Scientific Alfa Aesar
011305.22 |
100 g |
£526.62
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Erbium(III) chloride hydrate serves as a mild Lewis acid for selective acetalization. It combines with sodium borohydride and used as a selective reducing agent. It is also useful to form enolates from alfa-bromo ketones and oxyallyl cations from alfa,alfa'-dibromo ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19423-85-9 | |
273.61 | |
HDGGAKOVUDZYES-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[Er+3] |
Cl3Er | |
MFCD00010989 | |
erbium(3+) trichloride |
Specifications
19423-85-9 | |
Cl3Er | |
Soluble in water. | |
[Cl-].[Cl-].[Cl-].[Er+3] | |
273.61 | |
99.99% | |
Odorless | |
(REO) |
Crystalline Aggregates | |
MFCD00010989 | |
HDGGAKOVUDZYES-UHFFFAOYSA-K | |
erbium(3+) trichloride | |
273.62 (Anhydrous) | |
Reacton™ | |
Hygroscopic | |
Erbium(III) chloride hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 233-385-0
missing translation for 'rtecsNumber' : KD8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only