missing translation for 'onlineSavingsMsg'
Learn More
Erbium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11308617

Erbium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11308617
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11308617

Brand: Thermo Scientific Alfa Aesar 035739.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-41-7
Molecular Formula Cl3Er
Molecular Weight (g/mol) 273.609
MDL Number MFCD00010989
InChI Key HDGGAKOVUDZYES-UHFFFAOYSA-K
Synonym erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basisShow MoreShow Less
PubChem CID 66277
IUPAC Name trichloroerbium
SMILES Cl[Er](Cl)Cl

Specifications

Boiling Point 1500°C
Melting Point 774°C
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF.
Formula Weight 273.62
Percent Purity 99.9%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) chloride, Ultra dry
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06J067 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number035739.06,035739.14
Lot NumberW06J068 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number035739.06,035739.14
Lot NumberK18U006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number035739.06,035739.14
Lot NumberL06X005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number035739.06,035739.14
Lot NumberS30H088 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number035739.06,035739.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.