missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erbium(III) iodide, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals
£160.80 - £610.04
Chemical Identifiers
CAS | 13813-42-8 |
---|---|
Molecular Formula | ErI3 |
MDL Number | MFCD00016075 |
Synonym | erbium iii iodide, erbium iii ; iodide, acmc-1bohg, erbium iodide, |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11335637
|
Thermo Scientific Alfa Aesar
043372.09 |
10 g |
£160.80
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11345637
|
Thermo Scientific Alfa Aesar
043372.18 |
50 g |
£610.04
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13813-42-8 | |
ErI3 | |
MFCD00016075 | |
erbium iii iodide, erbium iii ; iodide, acmc-1bohg, erbium iodide, |
Specifications
1280°C | |
13813-42-8 | |
ErI3 | |
erbium iii iodide, erbium iii ; iodide, acmc-1bohg, erbium iodide, | |
99.99% | |
Hygroscopic | |
Erbium (III) iodide, Anhydrous |
1020°C | |
Powder | |
MFCD00016075 | |
547.97 | |
Reacton™ | |
(REO) |
Safety and Handling
missing translation for 'einecsNumber' : 237-471-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only