missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) oxalate decahydrate, REacton™, 99.99% (REO), Thermo Scientific™

£36.30 - £144.00

Chemical Identifiers

CAS 30618-31-6
Molecular Formula C6H26Er2O22
Molecular Weight (g/mol) 784.77
MDL Number MFCD00150142,MFCD00150142
InChI Key CYHKKMLSGVLNPD-UHFFFAOYSA-N
Synonym erbium oxalate decahydrate,
PubChem CID 131877482
IUPAC Name erbium;oxalic acid;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376668
SDS View Documents
Thermo Scientific Alfa Aesar
013216.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11386668
SDS View Documents
Thermo Scientific Alfa Aesar
013216.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium(III) oxalate decahydrate, REacton™, 99.99% (REO), Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.