missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) oxide, 99.8% (metals basis), Thermo Scientific Chemicals

For vacuum deposition

£155.78 - £457.28

Chemical Identifiers

CAS 12061-16-4
Molecular Formula Er2O3
Molecular Weight (g/mol) 382.52
MDL Number MFCD00010991
InChI Key ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
IUPAC Name dierbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Er+3].[Er+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327818
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
036372.14
25 g
£155.78
25g
Please to purchase this item. Need a web account? Register with us today!
11317818
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
036372.22
100 g
£457.28
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Erbium(III) oxide is used for vacuum deposition electrical, optical and photoluminescenc, gate dielectrics in semi conductor devices, and in Biomedicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12061-16-4
382.52
ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
[O--].[O--].[O--].[Er+3].[Er+3]
Er2O3
MFCD00010991
dierbium(3+) trioxidandiide

Specifications

2400°C
Sintered Lump
MFCD00010991
Soluble in water(0.0049g/L).
[O--].[O--].[O--].[Er+3].[Er+3]
382.52
99.8%
8.64 g/mL
Erbium(III) oxide
12061-16-4
Er2O3
14,3646
ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
dierbium(3+) trioxidandiide
382.56
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

missing translation for 'einecsNumber' : 235-045-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only