missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11301059
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Erbium trioxide is an important rare earth metal used in biomedicine. It is used as a gate dielectric in semi conductor devices since it has a high dielectric constant and a large band gap; as a burnable neutron poison for nuclear fuel and as coloring for glasses. Erbium oxide nanoparticles can be surface modified for distribution into aqueous and non-aqueous media for bioimaging due to its photoluminescence property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 12061-16-4
Molecular Formula Er2O3
Molecular Weight (g/mol) 382.52
MDL Number MFCD00010991
InChI Key ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
IUPAC Name dierbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Er+3].[Er+3]

Spécification

Melting Point 2400°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water.
Formula Weight 382.56
Sensitivity Hygroscopic
Appearance Powder or lump
Density 8.64
Chemical Name or Material Erbium(III) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'