missing translation for 'onlineSavingsMsg'
Learn More
Erbium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11321059

Erbium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11321059
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11321059

Brand: Thermo Scientific Alfa Aesar 011309.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Erbium trioxide is an important rare earth metal used in biomedicine. It is used as a gate dielectric in semi conductor devices since it has a high dielectric constant and a large band gap; as a burnable neutron poison for nuclear fuel and as coloring for glasses. Erbium oxide nanoparticles can be surface modified for distribution into aqueous and non-aqueous media for bioimaging due to its photoluminescence property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12061-16-4
Molecular Formula Er2O3
Molecular Weight (g/mol) 382.52
MDL Number MFCD00010991
InChI Key ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
IUPAC Name dierbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Er+3].[Er+3]

Specifications

Melting Point 2400°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water.
Formula Weight 382.56
Sensitivity Hygroscopic
Appearance Powder or lump
Density 8.64 g/cm3
Chemical Name or Material Erbium(III) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP22K013 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number011309.14,011309.36,011309.22,011309.06
Lot NumberW01J049 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number011309.14,011309.36,011309.22,011309.06
Lot NumberR17J016 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog Number011309.22
Lot NumberS29G011 Certificate TypeCertificate of Analysis Date26/05/2022 Catalog Number011309.14,011309.36,011309.22,011309.06
Lot NumberC11N18 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011309.14,011309.36,011309.22,011309.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.