missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog Number 11480301
Click to view available options
Quantity:
∼2 g
∼50 g
10 g
This item is not returnable. View return policy
14692-15-0
Cl3ErH12O18
573.687
MFCD00150232
JZRNHNTYRRYOCR-UHFFFAOYSA-K
erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade
20038200
erbium(3+);triperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]
This item is not returnable. View return policy
14692-15-0
Cl3ErH12O18
573.687
MFCD00150232
JZRNHNTYRRYOCR-UHFFFAOYSA-K
erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade
20038200
erbium(3+);triperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]

Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Pink
Physical Form Liquid
Quantity ∼2 g
Solubility Information Micible with water.
Formula Weight 573.70 (465.61 Anhydrous)
Grade Reagent
Odor Odorless
Appearance Pink liquid
Chemical Name or Material Erbium(III) perchlorate hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'