missing translation for 'onlineSavingsMsg'
Learn More
Erbium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11309237

Erbium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11309237
10 g
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11309237

Brand: Thermo Scientific Alfa Aesar 040092.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Erbium(III) sulfate octahydrate is used as a colorant in glass manufacturing and porcelain enamel glazes. High purity Erbium Sulfate is applied as dopant in making optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-52-4
MDL Number MFCD00149696
Synonym erbium sulfate octahydrate, dierbium trisulfate octahydrate, erbium sulfate er2 so4 3 octahydrate, erbium sulfate hydrate er2 so4 3.8h2o, sulfuric acid, erbium 3+ salt 3:2 , octahydrate, erbium sulfate, erbium sulfate-water 2/3/8, erbium iii sulfate octahydrate reo, dierbium 3+ ion octahydrate trisulfateShow MoreShow Less

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 766.82 (622.70 Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 3.205 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) sulfate octahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY15D014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040092.06,040092.18,040092.09
Lot NumberU23H064 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040092.06,040092.18,040092.09
Lot NumberB28L12 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040092.06,040092.18,040092.09
Lot NumberR03C043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040092.06,040092.18,040092.09
Lot NumberY15D013 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040092.06,040092.18,040092.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.