missing translation for 'onlineSavingsMsg'
Learn More

Erbium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 11393808
Click to view available options
Quantity:
10 mm
25 mm
50 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: Approx. 0.29g/mm, Cast rod, diameter is approximate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]

Especificaciones

Color Silver to Gray
Physical Form Cast Rod
Quantity 50 mm
Assay Percent Range 99.9% (Metals basis excluding Ta)
Odor Odorless
Packaging Mylar under Argon
Appearance Silver Gray rod
Density 9.066g/cm3
Chemical Name or Material Erbium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'