missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Erie Scientific™ Clipped-Corner Plain Glass Slides

Reduce the chances of cutting your finger or tearing a glove when you use these slides. Cole-Parmer™ Erie Scientific™ Clipped-Corner Plain Glass Slides feature clipped corners for easy handing, ideal for making smears.

Brand:  Cole-Parmer™ 48520-01

Product Code. 11785108

  • £88.85 / Pack of 144
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Reduce the chances of cutting your finger or tearing a glove when you use these slides. Cole-Parmer™ Erie Scientific™ Clipped-Corner Plain Glass Slides feature clipped corners for easy handing, ideal for making smears.

Drastically Clipped corners keep sample away from the edges of the slide.
  • Slides measure 75 x 25mm and are 1mm thick
  • Available in one gross (144 slides)
Specifications

Specifications

75 mm
Plain Glass
Microscope Slide
144 Pack
25 mm
1 mm
Clipped
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.