missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1 Monoclonal Antibody (ERK-6B11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 138600
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
MAPK3 (ERK1) is a serine/threonine kinase that is an important activator of p90, RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway. ERK1 is widely expressed and involved in the regulation of meiosis, mitosis, and post-mitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. Functionally, ERK1 is involved with a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. ERK1 is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms of ERK1 have been described.
Specifications
ERK1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide | |
P21708, P27361, Q63844 | |
MAPK3 | |
Synthetic Peptide corresponding to a section of the C-terminus of rat ERK1. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
ERK-6B11 | |
Unconjugated | |
MAPK3 | |
ERK; Erk1; ERK-1; Ert2; Esrk1; Extracellular signal-regulated kinase 1; extracellular signal-related kinase 1; HS44KDAP; HUMKER1A; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 3; MAP kinase isoform p44; MAP kinase2; MAPK 1; MAPK 3; MAPK1; MAPK2; MAPK3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase 3; mitogen-activated 3; mitogen-activated protein kinase 1; mitogen-activated protein kinase 3; MNK1; Mtap2k; p44; p44 MAP kinase; P44ERK1; p44-ERK1; P44MAPK; p44-MAPK; pp42/MAP kinase; PRKM3 | |
Mouse | |
Affinity chromatography | |
RUO | |
100855877, 26417, 50689, 5595 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ERK1 Monoclonal Antibody (ERK-6B11), Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction