Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03021422
Code : A0
Additional Details : Weight : 0.32400kg
 View more versions of this product
Product Code. 10303141
  • £71.29 / Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
DIN EN ISO 4797
25 mL
14/23 NS
Conical Ground Joint, Inscription Label
Borosilicate Glass, Duran Glass
42 mm