missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint

For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.

Brand:  Lenz Laborglasinstrumente™ 03021428

Code : NEW

Additional Details : Weight : 0.40000kg

 View more versions of this product

Product Code. 15300895

  • £73.83 / Pack of 10
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications

Specifications

DIN EN ISO 4797
50 mL
14/23 NS
Conical Ground Joint, Inscription Label
Borosilicate Glass, Duran Glass
51 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers