missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks, Narrow Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 07020037
Additional Details : Weight : 0.10000kg
Description
- Acc. to DIN ISO 1773
- With beaded rim
- With graduation
- Material: DURAN™
Specifications
22 mm | |
Duran Glass | |
10/Pk. |
100 mL | |
105 mm |