missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ ES 7X™-PF Cleaning Solution
GREENER_CHOICE

ES 7X™-PF Cleaning Solution is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.

Brand:  MP Biomedicals™ 097667194


 View more versions of this product

Product Code. 11409585

  • £205.00 / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Environmentally safe and phosphate free.
  • Developed to provide safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
  • Several convenient package sizes are available
  • Sold in concentrated form so the customer can prepare the appropriate dilution as needed
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid
Yellow
15°C to 30°C
4 Pk.
7 to 8
7X
Use on Glass, Plastic, benchtop equipment and tubing
1 gal.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ES-7X Cleaning Solution

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Dioctyl Sulfosuccinate Sodium Salt, 2-(2-butoxyethoxy)ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers