Learn More
MP Biomedicals™ ES 7X™-PF Cleaning Solution
ES 7X™-PF Cleaning Solution is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.
Brand: MP Biomedicals™ 097667193
Code : NEW
Additional Details : Weight : 3.79800kg
Description
- Environmentally safe and phosphate free.
- Developed to provide safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
- Several convenient package sizes are available
- Sold in concentrated form so the customer can prepare the appropriate dilution as needed
- Completely drains from glassware in seconds
- Eliminates residue leaving no interfering fluorescence
- pH neutral, and no need for pH adjustment at any concentration
- No etch to glass or plastic labware at any concentration
Specifications
Liquid | |
Yellow | |
15°C to 30°C | |
1 gal. |
7 to 8 | |
7X | |
Use on Glass, Plastic, benchtop equipment and tubing |
Safety and Handling
- ES-7X Cleaning Solution
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Dioctyl Sulfosuccinate Sodium Salt, 2-(2-butoxyethoxy)ethanol