missing translation for 'onlineSavingsMsg'
Learn More
Esculin hydrate, 97%, Thermo Scientific Chemicals Product Code.: 10146463

Esculin hydrate, 97%, Thermo Scientific Chemicals

Product Code. 10146463
5 g, Glass bottle
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10146463

Brand: Thermo Scientific Acros 117830050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-75-9
Molecular Formula C15H16O9
Molecular Weight (g/mol) 340.28
MDL Number MFCD00149492
InChI Key XHCADAYNFIFUHF-TYKRLAFXNA-N
Synonym esculin, aesculin, esculoside, --esculin, polychrome, 6,7-dihydroxycoumarin-6-o-glucoside, aesculinum, esculine, esculetin 6-o-glucoside, 6,7-dihydroxycoumarin 6-glucosideShow MoreShow Less
PubChem CID 5281417
ChEBI CHEBI:4853
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES OC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 203.0°C to 205.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 3753
Specific Rotation Condition - 86.00 (20.00°C c=2,dioxane/water1:1)
Specific Rotation − 86.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in hot water,soluble in hot alcohol,methanol,pyridine,,ethyl acetate and acetic acid
Formula Weight 340.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Esculin hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468360 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number117830050,117830500,117830100
Lot NumberA0462584 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number117830050,117830500,117830100
Lot NumberA0455887 Certificate TypeCertificate of Analysis Date29/08/2023 Catalog Number117830050,117830500,117830100
Lot NumberA0400668 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number117830050,117830500,117830100
Lot NumberA0409219 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number117830050,117830500,117830100
Product Identifier
  • Esculin hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Esculin hydrate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.