Learn More
Notrax ESD Heel Grounder
Click to view available options
Heel Grounder; For Use With: Connecting a Person with ESD Properties
Description
- A device for connecting a walking or standing person to ground by using a conductive rubber thread to provide the necessary electrical contact between the person and the floor mat
- Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
- Use these accessories to connect table work surface, employees and conductive material to a common grounding point
- This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
- For use in ESD working zones to protect electrostatic sensitive components
Specifications
Specifications
Product Type | Heel Grounder |
Material | Wire |
For Use With (Application) | Connecting a Person with ESD Properties |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Product Title
Notrax ESD Heel Grounder >
Spot an opportunity for improvement?Share a Content Correction