Learn More
Esonarimod, MedChemExpress
MedChemExpress Esonarimod (KE-298) is an antirheumatic agent.
Brand:  Medchem Express HY-19440
Encompass_Preferred
 View more versions of this product
Product Code. 16401819
  • POA / 1mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Esonarimod (KE-298) is an antirheumatic agent.
Specifications
Specifications
Esonarimod
101973-77-7
1 mg
C14H16O4S
KE-298
CC1=CC=C(C(CC(CSC(C)=O)C(O)=O)=O)C=C1
280.34
Research
Solid
White
Research
COVID-19-immunoregulation
DMSO : 100 mg/mL (356.71 mM; Need ultrasonic)
280.34
99.94%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Esonarimod, MedChemExpress