Learn More
Esonarimod, MedChemExpress
MedChemExpress Esonarimod (KE-298) is an antirheumatic agent.
Brand: Medchem Express HY-19440

Description
Esonarimod (KE-298) is an antirheumatic agent.Specifications
Esonarimod | |
101973-77-7 | |
50 mg | |
C14H16O4S | |
KE-298 | |
CC1=CC=C(C(CC(CSC(C)=O)C(O)=O)=O)C=C1 | |
280.34 | |
Research |
Solid | |
White | |
Research | |
COVID-19-immunoregulation | |
DMSO : 100 mg/mL (356.71 mM; Need ultrasonic) | |
280.34 | |
99.94% |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only