missing translation for 'onlineSavingsMsg'
Learn More

α-Estradiol, Tocris Bioscience™

Product Code. 18752764
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Endogenous estrogen receptor agonist

Endogenous estrogen receptor ligand (Ki values are 0.2 and 1.2 nM for ERα and ERβ receptors respectively).

β-Estradiol also available.

Specifications

CAS 57-91-0
Molecular Formula C18H24O2
Quantity 50 mg
Synonym alpha-estradiol, 17alpha-estradiol, alfatradiol, epiestradiol, 17-alpha-estradiol, 3,17-dihydroxyestratriene, 17a-estradiol, estradiol-17alpha, epiestrol, 17-epiestradiol
InChI Key VOXZDWNPVJITMN-SFFUCWETSA-N
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O
IUPAC Name (8R,9S,13S,14S,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
Molecular Weight (g/mol) 272.388
PubChem CID 68570
ChEBI CHEBI:17160
Formula Weight 272.38
Percent Purity >96%
Chemical Name or Material α-Estradiol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'