missing translation for 'onlineSavingsMsg'
Learn More

Estriol, MedChemExpress

MedChemExpress Estriol is an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells.

£75.02 - £105.27

Specifications

Chemical Name or Material Estriol
CAS 50-27-1
Color White
Purity Grade Notes Research
Molecular Formula C18H24O3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16498286
Encompass_Preferred
View Documents
Medchem Express
HY-B0412
100 mg
£75.02
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16432126
Encompass_Preferred
View Documents
Medchem Express
HY-B0412
500 mg
£105.27
500mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Estriol is an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells.
Specifications

Specifications

Estriol
White
C18H24O3
DMSO : 250 mg/mL (866.91 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.47 mM; ultrasonic and warming and heat to 80°C)
288.38
98.0%
Solid
50-27-1
Research
Oestriol
C[C@@]1([C@H]2O)[C@](C[C@H]2O)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1
288.38
Research
Cancer-programmed cell death
Videos
Safety and Handling

Safety and Handling

H351∣H360∣H362

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.