missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanediol, Extra Pure, SLR, Fisher Chemical™
C2H6O2, CAS Number-107-21-1, ethylene alcohol, glycol alcohol, 2-hydroxyethanol, 1,2-ethanediol, glycol, fridex, monoethylene glycol, tescol, 1,2-dihydroxyethane, ethylene glycol, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885
£48.05 - £860.00
Chemical Identifiers
CAS | 107-21-1 |
---|---|
Molecular Formula | C2H6O2 |
Molecular Weight (g/mol) | 62.068 |
MDL Number | 2885 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
Synonym | ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol |
PubChem CID | 174 |
ChEBI | CHEBI:30742 |
IUPAC Name | ethane-1,2-diol |
SMILES | C(CO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10395700
|
Fisher Chemical
E/0350/PB08 |
500 mL | HDPE plastic bottle |
£48.05
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10122000
|
Fisher Chemical
E/0350/15 |
1 L | Amber glass bottle |
£66.70
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10274430
|
Fisher Chemical
E/0350/17 |
2.5 L | Amber glass bottle |
£130.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10346420
|
Fisher Chemical
E/0350/PB17 |
2.5 L | HDPE plastic bottle |
£135.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10000130
|
Fisher Chemical
E/0350/25 |
25 L | Plastic container |
£860.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C2H6O2, CAS Number-107-21-1, ethylene alcohol, glycol alcohol, 2-hydroxyethanol, 1,2-ethanediol, glycol, fridex, monoethylene glycol, tescol, 1,2-dihydroxyethane, ethylene glycol, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885
Chemical Identifiers
107-21-1 | |
62.068 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
174 | |
ethane-1,2-diol |
C2H6O2 | |
2885 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
CHEBI:30742 | |
C(CO)O |
Specifications
107-21-1 | |
Colorless | |
196°C | |
2885 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
174 | |
CHEBI:30742 | |
0.12mmHg at 20°C |
-13°C | |
5.5 to 7.5 | |
C2H6O2 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
C(CO)O | |
62.068 | |
2.14 | |
62.06g/mol | |
Viscous Liquid |