missing translation for 'onlineSavingsMsg'
Learn More
Ethano L, 96%, technica L, denaturated with 5% methano L, Thermo Scientific Chemicals Product Code.: 13257983

Ethano L, 96%, technica L, denaturated with 5% methano L, Thermo Scientific Chemicals

Product Code. 13257983
2.5 L
Click to view available options
Quantity:
1 L
2.5 L
5 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 13257983

Brand: Thermo Scientific Acros 444140025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -117.0°C
Density 0.8000 g/mL
Boiling Point 78.0°C
Quantity 2.5 L
Flash Point 12°C
Packaging Glass Bottle
Refractive Index 1.3610 to 1.3630
Assay Percent Range 96%
Specific Gravity 0.8
Linear Formula C2H5OH
Merck Index 14, 3761
Solubility Information Solubility aq. soln.: completely soluble.
Formula Weight 46.06
Grade Technical
Chemical Name or Material Ethanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462437 Certificate TypeCertificate of Analysis Date15/05/2024 Catalog Number444140025,444140050,444140010
Lot NumberA0449782 Certificate TypeCertificate of Analysis Date10/10/2023 Catalog Number444140025,444140050,444140010
Lot NumberA0434354 Certificate TypeCertificate of Analysis Date02/10/2021 Catalog Number444140025,444140050,444140010
Lot NumberA0426461 Certificate TypeCertificate of Analysis Date27/01/2021 Catalog Number444140025,444140050,444140010
Lot NumberA0425352 Certificate TypeCertificate of Analysis Date18/12/2020 Catalog Number444140025,444140050,444140010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethanol, 96% denaturated with 5% methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P308-IF exposed or concerned:.

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethano L, 96%, technica L, denaturated with 5% methano L, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.