missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, 99.8%, for Analysis, Absolute, Thermo Scientific Chemicals Product Code.: 11924051

Ethanol, 99.8%, for Analysis, Absolute, Thermo Scientific Chemicals

Product Code. 11924051
1 L
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 11924051

Brand: Thermo Scientific Acros 445730010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

99.8% Ethanol for Analysis in a glass bottle.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • 99.8% Ethanol for Analysis, Absolute
  • GHS P Statement: Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower. Wear eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. If eye irritation persists: Get medical advice/attention.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114.0°C
Density 0.7900 g/mL
Boiling Point 78.0°C
Quantity 1 L
Flash Point 12°C
Packaging Glass Bottle
Assay Percent Range 99.8%
Specific Gravity 0.79
Linear Formula CH3CH2OH
Viscosity 1.2 mPa.s (20°C)
Merck Index 15, 3814
Solubility Information Solubility aq. soln.: completely soluble.
Formula Weight 46.07
Grade Analytical
Chemical Name or Material Ethanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463212 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog Number445730025,445730010
Lot NumberA0460002 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number445730025,445730010
Lot NumberA0457594 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog Number445730025,445730010
Lot NumberA0457595 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog Number445730025,445730010
Lot NumberA0452869 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog Number445730025,445730010
compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, 99.8%, for Analysis, Absolute, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.