missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 100%, technical, denaturated with 5% methanol, Thermo Scientific Chemicals

Product Code. 13287983
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -117.0°C
CAS 64-17-5
Density 0.8000 g/mL
Boiling Point 78.0°C
Quantity 5 L
Flash Point 12°C
Packaging Plastic Drum
Refractive Index 1.3610 to 1.3630
Assay Percent Range 100%
Specific Gravity 0.8
Molecular Formula C2H6O
Linear Formula C2H5OH
Merck Index 13, 3796
Solubility Information Solubility aq. soln.: completely soluble.
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Formula Weight 46.06
Grade Technical
Chemical Name or Material Ethanol
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Ethanol, 100% denaturated with 5% methanol
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol

For Manufacturing and Laboratory Use. Not for beverage use

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'