missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 70%, technical, not denaturated, Thermo Scientific Chemicals
64.82 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -48.0°C |
CAS | 64-17-5 |
Boiling Point | 85.0°C to 90.0°C |
Quantity | 5 L |
Flash Point | 21°C |
Packaging | Plastic Drum |
Assay Percent Range | 70% |
Molecular Formula | C2H6O |
Linear Formula | CH3CH2OH |
Merck Index | 15, 3814 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Product Identifier
- Ethanol, 60%, technical
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
For Manufacturing and Laboratory Use. Not for beverage use
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'